6-Bromo-2-(4-isopropoxyphenyl)quinoline-4-carboxylic acid

Supplier: Matrix Scientific

027875-500MG
101825-026EA 146.15 USD
101825-026
6-Bromo-2-(4-isopropoxyphenyl)quinoline-4-carboxylic acid
6-Bromo-2-(4-isopropoxyphenyl)quinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₁₉H₁₆BrNO₃
MDL Number: MFCD01924222
CAS Number: 351001-10-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR