6-Bromo-2-(2-chlorophenyl)quinoline-4-carboxylic acid

Supplier: Matrix Scientific

027874-500MG
101825-024EA 146.15 USD
101825-024
6-Bromo-2-(2-chlorophenyl)quinoline-4-carboxylic acid
6-Bromo-2-(2-chlorophenyl)quinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₁₆H₉BrClNO₂
MDL Number: MFCD02056013
CAS Number: 445289-24-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR