6-Bromo-2-(3,4-dimethylphenyl)quinoline-4-carboxylic acid

Supplier: Matrix Scientific

027870-500MG
101825-016EA 146.15 USD
101825-016
6-Bromo-2-(3,4-dimethylphenyl)quinoline-4-carboxylic acid
6-Bromo-2-(3,4-dimethylphenyl)quinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₁₈H₁₄BrNO₂
MDL Number: MFCD01923840
CAS Number: 351155-45-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR