6-Bromo-2-(2-ethoxyphenyl)quinoline-4-carboxylic acid

Supplier: Matrix Scientific

027866-500MG
101825-008EA 146.15 USD
101825-008
6-Bromo-2-(2-ethoxyphenyl)quinoline-4-carboxylic acid
6-Bromo-2-(2-ethoxyphenyl)quinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₁₈H₁₄BrNO₃
MDL Number: MFCD02055955
CAS Number: 438531-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR