6-Bromo-2-(2,4-dimethoxyphenyl)quinoline-4-carboxylic acid

Supplier: Matrix Scientific

027863-500MG
101825-002EA 146.15 USD
101825-002
6-Bromo-2-(2,4-dimethoxyphenyl)quinoline-4-carboxylic acid
6-Bromo-2-(2,4-dimethoxyphenyl)quinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₁₈H₁₄BrNO₄
MDL Number: MFCD01812952
CAS Number: 351329-40-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR