6-Bromo-2-(5-methylthien-2-yl)quinoline-4-carboxylic acid

Supplier: Matrix Scientific

027862-500MG
101825-000EA 146.15 USD
101825-000
6-Bromo-2-(5-methylthien-2-yl)quinoline-4-carboxylic acid
6-Bromo-2-(5-methylthien-2-yl)quinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₁₅H₁₀BrNO₂S
MDL Number: MFCD01922711
CAS Number: 350998-05-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR