6-Bromo-2-(4-ethylphenyl)quinoline-4-carboxylic acid

Supplier: Matrix Scientific

027861-500MG
101824-998EA 146.15 USD
101824-998
6-Bromo-2-(4-ethylphenyl)quinoline-4-carboxylic acid
6-Bromo-2-(4-ethylphenyl)quinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₁₈H₁₄BrNO₂
MDL Number: MFCD01922728
CAS Number: 350998-45-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR