6-Bromo-2-(3-methoxyphenyl)quinoline-4-carboxylic acid

Supplier: Matrix Scientific

027859-500MG
101824-994EA 146.15 USD
101824-994
6-Bromo-2-(3-methoxyphenyl)quinoline-4-carboxylic acid
6-Bromo-2-(3-methoxyphenyl)quinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₁₇H₁₂BrNO₃
MDL Number: MFCD00845856
CAS Number: 351329-64-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR