2-(2,5-Dimethylphenyl)-3-methylquinoline-4-carboxylic acid

Supplier: Matrix Scientific

027857-500MG
101824-990EA 146.15 USD
101824-990
2-(2,5-Dimethylphenyl)-3-methylquinoline-4-carboxylic acid
2-(2,5-Dimethylphenyl)-3-methylquinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₁₉H₁₇NO₂
MDL Number: MFCD01993734
CAS Number: 350999-29-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR