2-(3-Chlorophenyl)-3-methylquinoline-4-carboxylic acid

Supplier: Matrix Scientific

027855-500MG
101824-986EA 146.15 USD
101824-986
2-(3-Chlorophenyl)-3-methylquinoline-4-carboxylic acid
2-(3-Chlorophenyl)-3-methylquinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₁₇H₁₂ClNO₂
MDL Number: MFCD01993648
CAS Number: 350997-46-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR