2-(4-Methoxyphenyl)-3-methylquinoline-4-carboxylic acid

Supplier: Matrix Scientific

027853-500MG
101824-982EA 146.15 USD
101824-982
2-(4-Methoxyphenyl)-3-methylquinoline-4-carboxylic acid
2-(4-Methoxyphenyl)-3-methylquinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₁₈H₁₅NO₃
MDL Number: MFCD01114914
CAS Number: 108717-24-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR