2-(5-Chlorothien-2-yl)quinoline-4-carboxylic acid

Supplier: Matrix Scientific

027849-500MG
101824-974EA 146.15 USD
101824-974
2-(5-Chlorothien-2-yl)quinoline-4-carboxylic acid
2-(5-Chlorothien-2-yl)quinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₁₄H₈ClNO₂S
MDL Number: MFCD01114913
CAS Number: 329222-93-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR