2-(5-Ethylthien-2-yl)quinoline-4-carboxylic acid

Supplier: Matrix Scientific

027848-500MG
101824-972EA 230.77 USD
101824-972
2-(5-Ethylthien-2-yl)quinoline-4-carboxylic acid
2-(5-Ethylthien-2-yl)quinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₁₆H₁₃NO₂S
MDL Number: MFCD01471526
CAS Number: 351983-35-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR