2-(5-Methylthien-2-yl)quinoline-4-carboxylic acid

Supplier: Matrix Scientific

027847-500MG
101824-970EA 230.77 USD
101824-970
2-(5-Methylthien-2-yl)quinoline-4-carboxylic acid
2-(5-Methylthien-2-yl)quinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₁₅H₁₁NO₂S
MDL Number: MFCD00994756
CAS Number: 73775-26-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR