2-(3,4-Dimethoxyphenyl)quinoline-4-carboxylic acid

Supplier: Matrix Scientific

027844-500MG
101824-964EA 146.15 USD
101824-964
2-(3,4-Dimethoxyphenyl)quinoline-4-carboxylic acid
2-(3,4-Dimethoxyphenyl)quinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₁₈H₁₅NO₄
MDL Number: MFCD00219544
CAS Number: 133676-16-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR