2-(3-Butoxyphenyl)quinoline-4-carboxylic acid

Supplier: Matrix Scientific

027843-500MG
101824-962EA 146.15 USD
101824-962
2-(3-Butoxyphenyl)quinoline-4-carboxylic acid
2-(3-Butoxyphenyl)quinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₂₀H₁₉NO₃
MDL Number: MFCD01993645
CAS Number: 350997-43-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR