2-(4-Isopropoxyphenyl)quinoline-4-carboxylic acid

Supplier: Matrix Scientific

027839-500MG
101824-954EA 146.15 USD
101824-954
2-(4-Isopropoxyphenyl)quinoline-4-carboxylic acid
2-(4-Isopropoxyphenyl)quinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₁₉H₁₇NO₃
MDL Number: MFCD01922089
CAS Number: 425373-14-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR