2-(4-Propoxyphenyl)quinoline-4-carboxylic acid

Supplier: Matrix Scientific

027837-500MG
101824-950EA 146.15 USD
101824-950
2-(4-Propoxyphenyl)quinoline-4-carboxylic acid
2-(4-Propoxyphenyl)quinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₁₉H₁₇NO₃
MDL Number: MFCD01922013
CAS Number: 51842-69-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR