2-(3-Ethoxyphenyl)quinoline-4-carboxylic acid

Supplier: Matrix Scientific

027834-500MG
101824-944EA 146.15 USD
101824-944
2-(3-Ethoxyphenyl)quinoline-4-carboxylic acid
2-(3-Ethoxyphenyl)quinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₁₈H₁₅NO₃
MDL Number: MFCD01921947
CAS Number: 350989-62-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR