2-(4-Isopropylphenyl)-3-methylquinoline-4-carboxylic acid

Supplier: Matrix Scientific

027826-500MG
101824-928EA 146.15 USD
101824-928
2-(4-Isopropylphenyl)-3-methylquinoline-4-carboxylic acid
2-(4-Isopropylphenyl)-3-methylquinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₂₀H₁₉NO₂
MDL Number: MFCD01993650
CAS Number: 350997-49-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR