2-(3-Isobutoxyphenyl)quinoline-4-carboxylic acid

Supplier: Matrix Scientific

027823-500MG
101824-922EA 146.15 USD
101824-922
2-(3-Isobutoxyphenyl)quinoline-4-carboxylic acid
2-(3-Isobutoxyphenyl)quinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₂₀H₁₉NO₃
MDL Number: MFCD01993647
CAS Number: 350997-45-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR