3-(2-Furyl)propan-1-amine

Supplier: Matrix Scientific

027817-500MG
101824-910EA 387.69 USD
101824-910
3-(2-Furyl)propan-1-amine
3-(2-Furyl)propan-1-amine

Hazard: Irritant

Formula: C₇H₁₁NO
MDL Number: MFCD06589771
CAS Number: 4428-38-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR