4-[2-(2-Fluorophenyl)ethyl]piperidine

Supplier: Matrix Scientific

027806-500MG
101824-888EA 387.69 USD
101824-888
4-[2-(2-Fluorophenyl)ethyl]piperidine
4-[2-(2-Fluorophenyl)ethyl]piperidine

Hazard: Irritant

Formula: C₁₃H₁₈FN
MDL Number: MFCD05022472
CAS Number: 614731-85-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR