2,4-Dimethoxy-3-methyl-5-nitroaniline
Supplier: Matrix Scientific
027797-500MG
101824-872EA
290.77
USD
101824-872
2,4-Dimethoxy-3-methyl-5-nitroaniline
2,4-Dimethoxy-3-methyl-5-nitroaniline
Hazard: Irritant
Formula:
C₉H₁₂N₂O₄ |
MDL Number:
MFCD05858391 CAS Number: 704877-30-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...