4-(3-Bromophenyl)-1,3-thiazol-2-amine

Supplier: Matrix Scientific

Synonyms: 2-Amino-4-(3-bromophenyl)thiazole4-(3-Bromophenyl)thiazol-2-amine

027785-500MG
101824-848EA 76.92 USD
101824-848
4-(3-Bromophenyl)-1,3-thiazol-2-amine
4-(3-Bromophenyl)-1,3-thiazol-2-amine

Hazard: Irritant

Formula: C₉H₇BrN₂S
MW: 255.14 g/mol
MDL Number: MFCD00170238
CAS Number: 105512-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR