4-(2-Chlorophenyl)-1,3-thiazol-2-amine

Supplier: Matrix Scientific

Synonyms: 4-(2-Chlorophenyl)thiazol-2-amine

027783-500MG
101824-844EA 193.85 USD
101824-844
4-(2-Chlorophenyl)-1,3-thiazol-2-amine
4-(2-Chlorophenyl)-1,3-thiazol-2-amine

Hazard: Irritant

Formula: C₉H₇ClN₂S
MW: 210.69 g/mol
MDL Number: MFCD01315352
CAS Number: 21344-90-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR