1-(2-Methoxybenzyl)piperazine

Supplier: Matrix Scientific

027770-500MG
101824-822EA 100 USD
101824-822
1-(2-Methoxybenzyl)piperazine
1-(2-Methoxybenzyl)piperazine

Hazard: Irritant

Formula: C₁₂H₁₈N₂O
MDL Number: MFCD01860845
CAS Number: 55037-81-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR