N-[(1-Methyl-1H-pyrazol-4-yl)methyl]cyclohexanamine

Supplier: Matrix Scientific

027759-500MG
101824-812EA 193.85 USD
101824-812
N-[(1-Methyl-1H-pyrazol-4-yl)methyl]cyclohexanamine
N-[(1-Methyl-1H-pyrazol-4-yl)methyl]cyclohexanamine

Hazard: Irritant

Formula: C₁₁H₁₉N₃
MW: 193.29 g/mol
MDL Number: MFCD08691449
CAS Number: 1049983-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR