Methyl-5-(2-fluorophenyl)-4-(hydroxymethyl)-2-methylpyrrolidine-2-carboxylate

Supplier: Matrix Scientific

027753-500MG
101824-802EA 387.69 USD
101824-802
Methyl-5-(2-fluorophenyl)-4-(hydroxymethyl)-2-methylpyrrolidine-2-carboxylate
Methyl-5-(2-fluorophenyl)-4-(hydroxymethyl)-2-methylpyrrolidine-2-carboxylate

Hazard: Irritant

Formula: C₁₄H₁₈FNO₃
MDL Number: MFCD09971911
CAS Number: 1217644-56-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR