Methyl-5-(2-fluoro-5-methoxyphenyl)-4-(hydroxymethyl)-2-methylpyrrolidine-2-carboxylate

Supplier: Matrix Scientific

027749-500MG
101824-794EA 387.69 USD
101824-794
Methyl-5-(2-fluoro-5-methoxyphenyl)-4-(hydroxymethyl)-2-methylpyrrolidine-2-carboxylate
Methyl-5-(2-fluoro-5-methoxyphenyl)-4-(hydroxymethyl)-2-methylpyrrolidine-2-carboxylate

Hazard: Irritant

Formula: C₁₅H₂₀FNO₄
MDL Number: MFCD09971907
CAS Number: 1217831-96-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR