(6-Methoxy-2,3,4,9-tetrahydro-1H-carbazol-1-yl)methylamine

Supplier: Matrix Scientific

027739-500MG
101824-774EA 387.69 USD
101824-774
(6-Methoxy-2,3,4,9-tetrahydro-1H-carbazol-1-yl)methylamine
(6-Methoxy-2,3,4,9-tetrahydro-1H-carbazol-1-yl)methylamine

Hazard: Irritant

Formula: C₁₄H₁₈N₂O
MDL Number: MFCD09971901
CAS Number: 50845-44-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR