2-(4-Amino-3-oxobutyl)-1H-isoindole-1,3(2H)-dione hydrochloride

Supplier: Matrix Scientific

027737-500MG
101824-770EA 590.77 USD
101824-770
2-(4-Amino-3-oxobutyl)-1H-isoindole-1,3(2H)-dione hydrochloride
2-(4-Amino-3-oxobutyl)-1H-isoindole-1,3(2H)-dione hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₃ClN₂O₃
Melting Pt: 246 °C
MDL Number: MFCD26142754
CAS Number: 3330-06-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR