2-Ethyl-7-nitro-1,2,3,4-tetrahydroisoquinoline

Supplier: Matrix Scientific

027730-500MG
101824-756EA 290.77 USD
101824-756
2-Ethyl-7-nitro-1,2,3,4-tetrahydroisoquinoline
2-Ethyl-7-nitro-1,2,3,4-tetrahydroisoquinoline

Hazard: Irritant

Formula: C₁₁H₁₄N₂O₂
MDL Number: MFCD09971898
CAS Number: 711601-13-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR