2,1,3-Benzothiadiazol-4-ylamine

Supplier: Matrix Scientific

Synonyms: Benzo[c][1,2,5]thiadiazol-4-amine4-Aminobenzo-2,1,3-thiadiazole

027723-500MG
101824-742EA 217.35 USD
101824-742
2,1,3-Benzothiadiazol-4-ylamine
2,1,3-Benzothiadiazol-4-ylamine

Hazard: Irritant

Formula: C₆H₅N₃S
MW: 151.19 g/mol
MDL Number: MFCD00005810
CAS Number: 767-64-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR