2-(2-Chloroethyl)-6-methyl-1H-benzimidazole

Supplier: Matrix Scientific

027718-500MG
101824-732EA 290.77 USD
101824-732
2-(2-Chloroethyl)-6-methyl-1H-benzimidazole
2-(2-Chloroethyl)-6-methyl-1H-benzimidazole

Hazard: Irritant

Formula: C₁₀H₁₁ClN₂
MDL Number: MFCD08691513
CAS Number: 915921-57-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR