3-(2-Ethylpiperidin-1-yl)propanoic acid hydrochloride

Supplier: Matrix Scientific

027716-500MG
101824-728EA 290.77 USD
101824-728
3-(2-Ethylpiperidin-1-yl)propanoic acid hydrochloride
3-(2-Ethylpiperidin-1-yl)propanoic acid hydrochloride

Hazard: Irritant

Formula: C₁₀H₂₀ClNO₂
MDL Number: MFCD12026514
CAS Number: 933719-77-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR