(3,5-Dimethylpiperidin-1-yl)acetic acid hydrochloride

Supplier: Matrix Scientific

027712-500MG
101824-720EA 461.54 USD
101824-720
(3,5-Dimethylpiperidin-1-yl)acetic acid hydrochloride
(3,5-Dimethylpiperidin-1-yl)acetic acid hydrochloride

Hazard: Irritant

Formula: C₉H₁₈ClNO₂
MDL Number: MFCD14280123
CAS Number: 1158530-01-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR