7-Methyl-2-piperidin-3-yl-1H-benzimidazole

Supplier: Matrix Scientific

027698-500MG
101824-692EA 387.69 USD
101824-692
7-Methyl-2-piperidin-3-yl-1H-benzimidazole
7-Methyl-2-piperidin-3-yl-1H-benzimidazole

Hazard: Irritant

Formula: C₁₃H₁₇N₃
MDL Number: MFCD09971886
CAS Number: 933682-42-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR