6-Methoxy-2-piperidin-4-yl-1H-benzimidazole
Supplier: Matrix Scientific
027690-500MG
101824-676EA
387.69
USD
101824-676
6-Methoxy-2-piperidin-4-yl-1H-benzimidazole
6-Methoxy-2-piperidin-4-yl-1H-benzimidazole
Hazard: Irritant
Formula:
C₁₃H₁₇N₃O |
MDL Number:
MFCD09971881 CAS Number: 578709-04-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...