(6-Fluoro-1H-benzimidazol-2-yl)methanol

Supplier: Matrix Scientific

Synonyms: (5-Fluoro-1H-benzo[d]imidazol-2-yl)methanol

027688-500MG
101824-672EA 461.54 USD
101824-672
(6-Fluoro-1H-benzimidazol-2-yl)methanol
(6-Fluoro-1H-benzimidazol-2-yl)methanol

Hazard: Irritant

Formula: C₈H₇FN₂O
MW: 166.16 g/mol
MDL Number: MFCD09261475
CAS Number: 39811-07-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR