3-(5-Methoxy-1H-benzimidazol-2-yl)propan-1-amine dihydrochloride
Supplier: Matrix Scientific
027680-500MG
101824-656EA
461.54
USD
101824-656
3-(5-Methoxy-1H-benzimidazol-2-yl)propan-1-amine dihydrochloride
3-(5-Methoxy-1H-benzimidazol-2-yl)propan-1-amine dihydrochloride
Hazard: Irritant
Formula:
C₁₁H₁₇Cl₂N₃O |
MDL Number:
MFCD09029273 CAS Number: 897601-78-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...