3-(4,5-Dimethyl-1H-benzimidazol-2-yl)propan-1-amine

Supplier: Matrix Scientific

027678-500MG
101824-652EA 387.69 USD
101824-652
3-(4,5-Dimethyl-1H-benzimidazol-2-yl)propan-1-amine
3-(4,5-Dimethyl-1H-benzimidazol-2-yl)propan-1-amine

Hazard: Irritant

Formula: C₁₂H₁₇N₃
MW: 203.29 g/mol
MDL Number: MFCD09971875
CAS Number: 933719-67-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR