4-(4,5-Dimethyl-1H-benzimidazol-2-yl)butanoic acid

Supplier: Matrix Scientific

027673-500MG
101824-644EA 387.69 USD
101824-644
4-(4,5-Dimethyl-1H-benzimidazol-2-yl)butanoic acid
4-(4,5-Dimethyl-1H-benzimidazol-2-yl)butanoic acid

Hazard: Irritant

Formula: C₁₃H₁₆N₂O₂
MDL Number: MFCD09971873
CAS Number: 933682-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR