[(5-Methyl-1H-benzimidazol-2-yl)methoxy]acetic acid

Supplier: Matrix Scientific

027672-500MG
101824-642EA 746.15 USD
101824-642
[(5-Methyl-1H-benzimidazol-2-yl)methoxy]acetic acid
[(5-Methyl-1H-benzimidazol-2-yl)methoxy]acetic acid

Hazard: Irritant

Formula: C₁₁H₁₂N₂O₃
MDL Number: MFCD09971872
CAS Number: 876716-55-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR