3-(2,6-Dimethylpiperidin-1-yl)propan-1-ol

Supplier: Matrix Scientific

027664-500MG
101824-626EA 461.54 USD
101824-626
3-(2,6-Dimethylpiperidin-1-yl)propan-1-ol
3-(2,6-Dimethylpiperidin-1-yl)propan-1-ol

Hazard: Irritant

Formula: C₁₀H₂₁NO
MDL Number: MFCD06637419
CAS Number: 79156-90-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR