N-[2-(2-Methylpiperidin-1-yl)ethyl]propan-2-amine

Supplier: Matrix Scientific

027662-500MG
101824-622EA 387.69 USD
101824-622
N-[2-(2-Methylpiperidin-1-yl)ethyl]propan-2-amine
N-[2-(2-Methylpiperidin-1-yl)ethyl]propan-2-amine

Hazard: Irritant

Formula: C₁₁H₂₄N₂
MDL Number: MFCD09971867
CAS Number: 933744-98-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR