N-Ethyl-N-[2-(2-ethylpiperidin-1-yl)ethyl]amine

Supplier: Matrix Scientific

027658-500MG
101824-614EA 290.77 USD
101824-614
N-Ethyl-N-[2-(2-ethylpiperidin-1-yl)ethyl]amine
N-Ethyl-N-[2-(2-ethylpiperidin-1-yl)ethyl]amine

Hazard: Irritant

Formula: C₁₁H₂₄N₂
MDL Number: MFCD09971864
CAS Number: 933738-32-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR