N-[(5-Ethyl-1,3,4-oxadiazol-2-yl)methyl]-N-methylamine

Supplier: Matrix Scientific

027648-500MG
101824-594EA 387.69 USD
101824-594
N-[(5-Ethyl-1,3,4-oxadiazol-2-yl)methyl]-N-methylamine
N-[(5-Ethyl-1,3,4-oxadiazol-2-yl)methyl]-N-methylamine

Hazard: Irritant

Formula: C₆H₁₁N₃O
MDL Number: MFCD08691563
CAS Number: 915920-74-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR