2-(2,6-Dimethylmorpholin-4-yl)ethanamine

Supplier: Matrix Scientific

027647-500MG
101824-592EA 76.92 USD
101824-592
2-(2,6-Dimethylmorpholin-4-yl)ethanamine
2-(2,6-Dimethylmorpholin-4-yl)ethanamine

Hazard: Irritant

Formula: C₈H₁₈N₂O
MDL Number: MFCD08444750
CAS Number: 244789-18-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR