2-(7-Methyl-1H-benzimidazol-2-yl)ethanol

Supplier: Matrix Scientific

027646-500MG
101824-590EA 290.77 USD
101824-590
2-(7-Methyl-1H-benzimidazol-2-yl)ethanol
2-(7-Methyl-1H-benzimidazol-2-yl)ethanol

Hazard: Irritant

Formula: C₁₀H₁₂N₂O
MDL Number: MFCD08691508
CAS Number: 915921-55-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR